missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ 0.2 mL PCR Tubes with Attached Caps, BIO-CERT™ PCR QUALITY

Thin Wall PCR Tubes with attached caps ensure precise thermal transfer, RNase-/DNase-/DNA-free, and manufactured without lubricants or additives that could contaminate samples.

Supplier:  BRANDTECH™ BRAND™ 781304

 View more versions of this product

Catalog No. 1388257


Only null left

Description

Description

BRAND ™ Disposable PCR Tubes are designed to deliver exceptional performance for PCR setups, small- and medium-throughput applications, and other workflows requiring precise thermal transfer and high cleanliness. Made from high-quality virgin polypropylene, these tubes ensure reliable results with features that enhance usability and sample integrity. Key features include:

  • BIO-CERT™ PCR QUALITY: Certified free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors, ensuring the highest level of cleanliness for sensitive PCR applications.
  • Superior Heat Transfer: Extra thin-walled tubes optimize thermal transfer during PCR, reducing cycle times and providing consistent performance over a wide temperature range.
  • Reliable Sealing Caps: Tight-sealing caps prevent sample evaporation and loss due to heating, while offering easy-to-open and close functionality.
  • Color Options: Offered in multiple colors for simplified visual sample tracking and organization.
  • High-Quality Construction: Manufactured from high-quality virgin resins without lubricants or chemical additives that could contaminate reagents or samples.
  • Autoclavable: Designed to withstand autoclaving at 121°C (2 bar) in compliance with DIN EN 285.
Specifications

Chemical Identifiers

RNase, DNase and DNA free
1000 Pack
Polypropylene
Yes
Tube
Thin Wall PCR Tube
0.2 mL
Non-sterile
Autoclavable
Single-use
0.2 mL

Specifications

RNase, DNase and DNA free
0.2 mL
Thermal cyclers for excellent reproducible results in most PCR applications
Polypropylene
Autoclavable
Single-use
0.2 mL
Blue
1000 Pack
Non-sterile
Flat Cap
Yes
Tube
Thin Wall PCR Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions