missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ Cuvette Caps
Description
- Round cuvette caps
- Fits ultra-micro cuvettes
- Provide tight seal
- Allows storage of samples to -20°
Specifications
Specifications
| Type | Cuvette Cap |
| Color | Blue |
| Disposable | Single-use |
| Material | Polyethylene |
| Quantity | 100 Pack |
| For Use With (Equipment) | Ultramicro UV Cuvette |
| Shape | Round |