missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ VACUUBRAND™ Collection Vessles and Adapter Kit for BVC Fluid Aspiration Systems

Replacement parts and accessories for VACUUBRAND fluid aspiration systems

$545.62 - $2891.44

Specifications

For Use With BVC fluid aspiration system
View More Specs

Products 3
Catalog Number Mfr. No. Type Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Type Capacity (Metric) Price Quantity  
13-688-015
View Documents
BrandTech™ VACUUBRAND™
20635810
Collection Vessel 4 L
Each for $2,891.44
Only null left
 
13-688-016
View Documents
BrandTech™ VACUUBRAND™
20635809
Collection Vessel 2 L
Each for $2,767.56
Only null left
 
13-688-017
View Documents
BrandTech™ VACUUBRAND™
20635839
Adapter
Each for $545.62
Only null left
 
Description

Description

  • The BVC family of fluid aspiration systems delivers compact and quiet solutions for cell culture work.
  • They provide the flexibility to accommodate decontamination protocol whether autoclaving or disinfecting in the bottle.Collection Vessel
  • Collection vessel: available in 2L and 4L capacities
  • Adapter Kit: to fit 2 liter glass collection vessel to BVC control and professional models
Specifications

Specifications

BVC fluid aspiration system
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'