missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ S 96-well Microplates
Nontreated sterile microplates for diverse applications
Description
- PS surface
- Medium binding
- Sterilized via beta-radiation
- Standard plates available in U-, V-, F-, C-shapes
- Transparent bottom available in F- shape
- With lid
- Available in clear, white or black
- Gray alphanumeric embossing on white and black solid plates for easy well identification
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Bacteriological assays, screenings, storage.
Specifications
| Includes Barcode | |
| Standard | |
| 8 x 12 | |
| Medium Binding | |
| 96-well Assay Microplate | |
| 330 μL | |
| Sterile | |
| Flat Bottom (F-well) | |
| 50/Cs. |
| ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS | |
| With Lid | |
| Individually Wrapped | |
| Black, Clear Bottom | |
| Polystyrene | |
| 96 | |
| Non-treated | |
| Bacteriological assays, screening and for storage |