Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ S 96-well Microplates
Nontreated sterile microplates for diverse applications
 View more versions of this product
Catalog No. 13-882-213
Only null left
Explore available promotions
Description
Description
  • PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottom available in F- shape
  • With lid
  • Available in clear, white or black
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Bacteriological assays, screenings, storage.
Specifications
Specifications
Includes Barcode
Standard
8 x 12
Clear
350 μL
Sterile
Rounded Square
50/Cs.
ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS
With Lid
Medium Binding
Polystyrene
96
Non-treated
Bacteriological assays, screening and for storage
Product Suggestions
Product Suggestions