missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ 1536-Well Microplates
Non-treated microplates for diverse applications
Description
- Nontreated PS surface
- Medium binding, nonsterile
- Standard plate for most applications
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Homogenous assays, screening, and storage.
Chemical Identifiers
| No Barcode | |
| ISO 11137, AAMI | |
| Without Lid | |
| Not Autoclavable | |
| Single-use | |
| Polystyrene | |
| 1536 | |
| Non-sterile | |
| Sterile applications, bacteriological assays |
| Medium Binding | |
| Standard | |
| 32 x 48 | |
| Untreated | |
| Full | |
| 10 μL | |
| 10 μL | |
| Non-treated | |
| 50/Cs. |
Specifications
| No Barcode | |
| ISO 11137, AAMI | |
| Without Lid | |
| Not Autoclavable | |
| Clear | |
| 1536-Well Cell Culture Multiwell Plates | |
| Full | |
| 10 μL | |
| 10 μL | |
| Non-treated | |
| Sterile applications, bacteriological assays |
| Medium Binding | |
| Standard | |
| 32 x 48 | |
| Untreated | |
| Single-use | |
| Plate Readers | |
| Polystyrene | |
| 1536 | |
| Non-sterile | |
| Square, Flat Bottom (F-well) | |
| 50/Cs. |