missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Real-Time PCR (qPCR) 96-Well Microplates, BIO-CERT™ PCR QUALITY, Low Profile

Catalog No. p-4484638
Change view
Click to view available options
Includes:
Plates and polyester seals
Plates only
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
13882330 Plates only
13882331 Plates and polyester seals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13882330 Supplier BRANDTECH™ BRAND™ Supplier No. 781364
Only null left

Optimized for accurate qPCR results, these plates are fully compatible with Roche™ LightCycler™ Systems.

Designed for optimal performance in fluorescence signal analysis and DNA quantification, these plates are ideal for qPCR applications. Key features include:

  • Enhanced Signal Reflection: White surface and titanium dioxide (TiO2) coloring maximize fluorescence signal reflection for precise quantification.
  • Exceptional Purity: BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors.
  • Advanced Manufacturing Standards: Produced in state-of-the-art clean room conditions for consistent quality.
  • Superior Thermal Performance: Extra-thin walls provide efficient thermal transfer.
  • User-Friendly Design:
    • Smooth surfaces optimize signal reflection
    • Blue embossed alphanumeric coding and a cut corner design ensure easy orientation
    • Low-profile, semi-skirted frame with a non-elevated rim offers compatibility with various thermal cyclers, including Roche™ LightCycler™ systems
  • Additional Convenience: Part no. 781365 includes 50 sheets of qPCR polyester sealing film

Specifications

Barcode No Barcode
Binding Property Untreated
Certifications/Compliance DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging Bagged
Color White
Disposable Single-use
Product Type qPCR 96-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Semi-skirted
Material Polypropylene
Volume (Metric) Well 0.15 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
Includes Plates only
Profile Low
Quantity 50/Pk.
RNase Free Yes
Show More Show Less