Learn More
BRANDTECH™ BRAND™ 96-Well Skirted Microplates, BIO-CERT™ PCR QUALITY, Low Profile
Optimized for reliable and precise results in a wide range of PCR applications.
Supplier:  BRANDTECH™ BRAND™ 781377
 View more versions of this product
Catalog No. 14-388-165
Only null left
Explore available promotions
Description
Description
BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
  • Exceptional Quality and Purity
    • BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
    • Free of contaminants that could interfere with reagents and samples
    • Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
  • Precision Thermal Performance
    • Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
    • Designed for rapid and accurate heat transfer, reducing cycle times
  • Wide Compatibility
    • Suitable for most commercially available thermal cyclers
    • White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
  • User-Friendly Features
    • Blue alphanumeric coding for easy sample identification
    • Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
    • Skirted plates provide extra rigidity, ideal for automated pipetting systems and transport
Specifications
Specifications
No Barcode
DNA, DNase, RNase-free
8 x 12
Bagged
Single-use
Thermal Cyclers
Polypropylene
96
Non-treated
Yes
Plates only
Low
Untreated
Standard
Autoclavable
Clear
PCR 96-well plate
Fully skirted
0.15 mL
Non-sterile
Conical Bottom (V-well)
qPCR
50
Yes
Product Suggestions
Product Suggestions