missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ 96-Well Semi-Skirted Microplates, BIO-CERT™ PCR QUALITY, Low Profile
Optimized for reliable and precise results in a wide range of PCR applications.
$941.06 - $1051.00
Specifications
| Barcode | No Barcode |
|---|---|
| Binding Property | Untreated |
| Certifications/Compliance | DNA, DNase, RNase-free |
| Format | Standard |
| Lid | Without Lid |
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|
14-388-162
|
BRANDTECH™ BRAND™
781371 |
Clear |
Pack of 50 Each for $941.06
|
|
|||||
|
14-388-168
|
BRANDTECH™ BRAND™
781372 |
White |
Pack of 50 Each for $1,051.00
|
|
|||||
Description
BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
- Exceptional Quality and Purity
- BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
- Free of contaminants that could interfere with reagents and samples
- Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
- Precision Thermal Performance
- Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
- Designed for rapid and accurate heat transfer, reducing cycle times
- Wide Compatibility
- Suitable for most commercially available thermal cyclers
- White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
- Skirted plates provide extra rigidity, ideal for automated pipetting systems and transport
- User-Friendly Features
- Blue alphanumeric coding for easy sample identification
- Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
Specifications
| No Barcode | |
| DNA, DNase, RNase-free | |
| Without Lid | |
| Autoclavable | |
| Untreated | |
| Thermal Cyclers | |
| Polypropylene | |
| 96 | |
| Non-sterile | |
| 14.4 mL | |
| Yes | |
| Plates only | |
| Low |
| Untreated | |
| Standard | |
| 8 x 12 | |
| 10 bags of 5 | |
| Single-use | |
| Semi-skirted | |
| 0.15 mL | |
| 0.15 mL | |
| Non-treated | |
| Conical Bottom (V-well) | |
| qPCR | |
| 50 | |
| Yes |