missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Pipet Tips, 5 to 300 μL

Catalog No. 13889144
Change view
Click to view available options
Tip Style:
Standard
Format:
Bulk
Rack
Refill
Packaging:
Bulk, 10,000 pieces
Bulk, 1000 pieces
TipBox, 5 xoxes of 96 pieces
TipRack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style Format Packaging
13889144 Standard Bulk Bulk, 1000 pieces
13889150 Standard Bulk Bulk, 10,000 pieces
13889156 Standard Refill TipRack
13889175 Standard Rack TipBox, 5 xoxes of 96 pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13889144 Supplier BRANDTECH™ BRAND™ Supplier No. 732010
Only null left

Designed for high-quality micro-pipettes and suitable for various applications. BRAND™ 5 to 300 μL pipette tips ensure precise and efficient pipetting. Tips are cleanroom-produced, free from contaminants, and environmentally friendly.

Used for virtually all pipettes with yellow color code.

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Nonsterile
  • All unopened packages are DNA-, RNAse, ATP- and endotoxin-free
  • Thin-walled for high translucency, easy ejection and low waste
  • Graduation at 50, 100 and 300 μL for quick volume check
  • Available in either standard or ultra low retention types
  • Available either in bulk packages or in convenient, ecological rack format
  • Racked tips are colorless and placed into green-colored tip tray

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and minimal sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can increase sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties

Specifications

Description Life Science Quality
Volume (Metric) 5 to 300 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Length (English) 2.08 in.
Length (Metric) 5.3 cm
Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) Reverse pipetting, plate washing, working with multichannel pipettes
Format Bulk
Tip Style Standard
Packaging Bulk, 1000 pieces
Product Type Pipette Tips
Disposable Limited-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Graduations 50, 100, 300 μL
Show More Show Less