missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Pipette Tips, 0.5 to 20 μL

Catalog No. 13889141
Change view
Click to view available options
Packaging:
Bulk
TipBox
TipRack
Quantity:
10,000/Pk.
1000/Pk.
480/Pk.
960/Pk.
Filtered:
Non-filtered
Tip Style:
Narrow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity Filtered Tip Style
13889141 Bulk 1000/Pk. Non-filtered Narrow
13889147 Bulk 10,000/Pk. Non-filtered Narrow
13889153 TipRack 960/Pk. Non-filtered Narrow
13889172 TipBox 480/Pk. Non-filtered Narrow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13889141 Supplier BRANDTECH™ BRAND™ Supplier No. 732004

Manufactured from high-purity, oleamide and diHEMDA-free polypropylene. Pipet Tips, 0.5 to 20μL allow pipetting in microtubes and microplates without touching the walls. Gradations marked at 2μL and 10μL provide quick volume assessment.

  • Nonsterile , high-purity polypropylene is free from oleamide and diHEMDA.
  • Gradations marked at 2 μL and 10 μL provide quick volume assessment.
  • Slender design allows pipetting in microtubes and microplates without touching the walls.
  • Available in either standard or ultra low retention types.
  • Available either in bulk packages or in convenient rack-and-stack formats.

Ultra low retention tips

  • Produced through special, patented physicochemical process.
  • Homogeneous, defect-free surface thus produced has low surface tension.
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media.
  • For biological samples that contain detergents such as Triton™ X-100, SDS, Tween, etc.
  • High chemical resistance.
  • Tips can be autoclaved at 121°C (2 bar) without damaging material properties.

Specifications

Description Life Science Quality
Volume (Metric) 0.5 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Length (English) 1.8 in.
Length (Metric) 46 mm
Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) Reverse pipetting, plate washing, working with multichannel pipettes
Format Bulk
Tip Style Narrow
Packaging Bulk
Quantity 1000/Pk.
Product Type Pipette Tips
Disposable Limited-use
For Use With (Equipment) Universal Fit Ultramicro
Graduated Yes
Material Polypropylene
Fit for Purpose PCR Ready
Graduations 2, 10 μL
Show More Show Less