missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Self-adhesive Plate Sealing Film

Catalog No. 1388309
Change view
Click to view available options
Material:
Aluminum
Polyester
Polyethylene, Polypropylene
Polypropylene
Rayon
For Use With (Application):
Automation
Cell and tissue culture
Cold storage
ELISA, PCR
Storage
No. per Pack:
100 Sheets
50 Sheets
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material For Use With (Application) No. per Pack
1388309 Polyester ELISA, PCR 100 Sheets
1388295 Rayon Cell and tissue culture 100 Sheets
14380807 Polypropylene Storage 100 Sheets
13882329 Aluminum Cold storage 100 Sheets
13878201 Rayon Cell and tissue culture 50 Sheets
14380809 Polyethylene, Polypropylene Automation 50 Sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 1388309 Supplier BRANDTECH™ BRAND™ Supplier No. 781390

Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.

  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl

Specifications

Color Clear
Material Polyester
Sterility Non-sterile
Autoclavable Not Autoclavable
No. per Pack 100 Sheets
Evaporation Barrier Yes
For Use With (Application) ELISA, PCR
Pre-slit No
Product Type Sealing Film
Adhesive Pressure Sensitive
Thickness 9.96 mil.
Pierceable No
Temperature Range (Metric) -40°C to +120°C
Show More Show Less