missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Deep Well Plates

Seamlessly integrates into multi-channel systems and automation workflows.

Supplier:  BRANDTECH™ BRAND™ 701350

 View more versions of this product

Catalog No. 13-882-52


Only null left

Description

Description

BRAND™ Deep Well Plates are designed for a broad range of applications, including sample storage, high-throughput screening (HTS), cell and tissue culture, serial dilutions, and reagent transfers. Most plates are manufactured from high-quality virgin polypropylene, with one highly transparent polystyrene variant for applications requiring optical clarity.

  • Available in 96- and 384-well formats
  • Manufactured from high-quality virgin polypropylene or polystyrene
  • BIO-CERT™ PCR Quality are free from human DNA, DNase, RNase, and pyrogens
  • Polypropylene plates are autoclavable, chemically resistant, and suitable for storage down to -80°C (-112°F)
  • Polystyrene plate offers excellent transparency and is suitable for storage down to -20°C (-4°F)
  • ANSI/SLAS 1 and 4 compliant for compatibility with automated systems
  • Alphanumeric well labeling and stackable design for efficient organization
  • Notched corners ensure correct orientation in robotic and automated liquid-handling systems
  • Compatible with sealing mats or adhesive films to minimize evaporation and contamination
Specifications

Specifications

No Barcode
8 x 12
Clear
Polypropylene
96
Non-treated
Flexible mat covers or adhesive sealing films to reduce sample evaporation and contamination
U-bottom
Standard
Autoclavable
Deep-well Plate
1.1 mL
Non-sterile
Round
24
24 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions