missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Deep Well Plates

Catalog No. 1388250
Change view
Click to view available options
Volume (Metric) Well:
0.3 mL
0.5 mL
1.1 mL
2.2 mL
Format:
Standard
Well Shape:
Round
Round Bottom (U-well)
Square, Conical Bottom (V-well)
Square, Round Bottom (U-well)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Well Format Well Shape
1388250 0.5 mL Standard Round Bottom (U-well)
1388251 1.1 mL Standard Round Bottom (U-well)
1388252 1.1 mL Standard Round
1388253 2.2 mL Standard Square, Round Bottom (U-well)
14380804 0.3 mL Standard Square, Conical Bottom (V-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 1388250 Supplier BRANDTECH™ BRAND™ Supplier No. 701346

Seamlessly integrates into multi-channel systems and automation workflows.

BRAND™ Deep Well Plates are designed for a broad range of applications, including sample storage, high-throughput screening (HTS), cell and tissue culture, serial dilutions, and reagent transfers. Most plates are manufactured from high-quality virgin polypropylene, with one highly transparent polystyrene variant for applications requiring optical clarity.

  • Available in 96- and 384-well formats
  • Manufactured from high-quality virgin polypropylene or polystyrene
  • BIO-CERT™ PCR Quality are free from human DNA, DNase, RNase, and pyrogens
  • Polypropylene plates are autoclavable, chemically resistant, and suitable for storage down to -80°C (-112°F)
  • Polystyrene plate offers excellent transparency and is suitable for storage down to -20°C (-4°F)
  • ANSI/SLAS 1 and 4 compliant for compatibility with automated systems
  • Alphanumeric well labeling and stackable design for efficient organization
  • Notched corners ensure correct orientation in robotic and automated liquid-handling systems
  • Compatible with sealing mats or adhesive films to minimize evaporation and contamination

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Color Clear
Product Type 96-well Assay Microplate
Material Polypropylene
Volume (Metric) Well 0.5 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
For Use With (Application) Flexible mat covers or adhesive sealing films to reduce sample evaporation and contamination
No. per Pack 48
Profile U-bottom
Quantity 48 Pk.
Show More Show Less