missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ BRAND™ Four-Clear Sided Disposable Cuvettes for Fluorescence
BRAND™ UV-transparent cuvettes are the cost-effective alternative to glass or quartz cuvettes, perfect for numerous applications.
$408.34 - $1917.31
Specifications
| Sterility | Non-sterile |
|---|---|
| Type | Fluorimetry Cuvette |
| Color | Transparent |
| Disposable | Single-use |
| Capacity (Metric) | 2.5 to 4.5 mL |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
14-381-602
|
BrandTech™ BRAND™
759125 |
100 |
Pack of 100 Each for $408.34
|
|
|||||
|
14-381-603
|
BrandTech™ BRAND™
759128 |
500 |
Pack of 500 Each for $1,917.31
|
|
|||||
Description
- BRAND™ premium quality four-sided clear disposable cuvettes are tailored for superior fluorescence applications
- Made of proprietary UV-plastic
- Optimal for assays in the 230 to 800 nm range, offering virtually no autofluorescence for an excellent signal-to-noise ratio
- Equipped with a 10 mm light path, these cuvettes are designed to fit most fluorimeters
Specifications
| Non-sterile | |
| Transparent | |
| 2.5 to 4.5 mL | |
| Cuvette only | |
| 230 to 800 nm | |
| flourometers | |
| Fluorimetry | |
| Rectangle |
| Fluorimetry Cuvette | |
| Single-use | |
| 12.5mm | |
| 10 mm | |
| 45 mm | |
| 12.5 mm | |
| 10 mm |