Learn More
Stemcell Technologies Inc BrainPhys™ Imaging Optimized Medium, Size: 500 mL
Catalog No. NC1991492
Encompass
BrainPhys™ Imaging Optimized Medium is a serum-free neuronal basal medium (based on the BrainPhys™ formulation published by Cedric Bardy and Fred H. Gage) that is optimized for imaging applications.Applications of BrainPhys™ Imaging Optimized Medium include live fluorescent imaging (calcium imaging and optogenetics) and neuronal culture. In addition to the removal of phenol red, the formulation has been modified to reduce background fluorescence and increase stability upon repeated exposure to light (M Zabolocki et al. Nat Comm, 2020).
Catalog No. NC1991492 Supplier Stemcell Technologies Inc Supplier No. STM05796
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BrainPhys™ Imaging Optimized Medium is a serum-free neuronal basal medium (based on the BrainPhys™ formulation published by Cedric Bardy and Fred H. Gage) that is optimized for imaging applications.Applications of BrainPhys™ Imaging Optimized Medium include live fluorescent imaging (calcium imaging and optogenetics) and neuronal culture. In addition to the removal of phenol red, the formulation has been modified to reduce background fluorescence and increase stability upon repeated exposure to light (M Zabolocki et al. Nat Comm, 2020).