missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brain Natriuretic Peptide (1-32), Human

For cellular and molecular biology applications

$692.00 - $692.00

Specifications

Species Human
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 3464.03
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66167MCR
View Documents
Thermo Scientific Chemicals
J66167MCR
1 mg
Each for $692.00
Only null left
 
Description

Description

Brain natriuretic peptide is originally isolated from brain, but is mainly produced in myoendocrine cells of the heart ventricles from which it is released into the circulation. It is involved in blood pressure control and cardiovascular homeostasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Human
-30°C to -10°C
3464.03
RUO - research use only
HPLC
Solid
Brain Natriuretic Peptide (1-32), human
BNP (1-32), human
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only