missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyPrinter S3700 Multicolor and Cut Sign and Label Printer with Color Label Materials Kit

Catalog No. 17377006
Change view
Click to view available options
Color:
Blue, Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17377006 Blue, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17377006 Supplier Brady™ Supplier No. S3700KITCOLOR
Only null left

Get all the colors and capabilities you need in this convenient Printer and Materials kit to create safety labels, Lean visuals, lockout tags, work-in-process labels and more that truly pop.

  • Printer optimized for 1-, 2- or 4-color panel ribbons for multicolor made easy
  • No calibrating, no threading, no waste (single-color ribbon only in kit)
  • Create shapes, arrows, letters and more with the built-in XY plotter cutter that slices straight lines, smoother curves and tighter connection points
  • Get your print jobs done right in less time with 20-second ribbon and material changeovers plus up to 5-in.-per-second print speed
  • Empower any user to print how they choose with time-saving onboard templates, intuitive PC labeling software and a mobile app
  • Simply tap, print, go with preprogrammed onboard label templates plus 600 barcodes and symbols
  • Durable, low-shrink All-weather Permanent Adhesive Vinyl Label Tape with ultra-aggressive adhesive is ideal for just about any industrial surface
  • Waste less, print more with print ribbons that last up to 60% longer by operating independently of label roll

Specifications

Product Type Label Printer Kit
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Thermal Transfer
Length (English) 14.25 in.
Width (English) 19 in.
Length (Metric) 361.95 mm
Width (Metric) 482.6 mm
Height (English) 11 in.
Height (Metric) 279.4 mm
Dimensions (L x W x H) 14.25 x 19 x 11 in.
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Multicolor
Includes S3700 Safety and Facility Printer, Brady Workstation Safety and Facility ID Software, Cleaning Kit, Mouse, Power Cord, Quick Start Guide, USB Cord, USB Flash Drive, 4 in. Continuous White, Blue, Green, Red, and Yellow Vinyl Labels, Black Ribbon Cartridge, White Ribbon Cartridge
Languages Indonesian, Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
Memory 512 MB
Model S3700
Print Speed 5 in./sec.
Resolution 300 dpi
Temperature Range (English) 59°F to 104°F
Temperature Range (Metric) 15°C to 40°C
For Use With (Application) Asset Tracking, Cold Temperature Labeling, Equipment Labeling, Electrical Labeling, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Food and Beverage Process Labeling, Healthcare and Hospital Labeling, Barcode Labeling
Warranty 5 Years
Power Supply Details AC Adaptor
Voltage 110/240 V
Keyboard Type QWERTY (US)
Show More Show Less