missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyPrinter i5100 i7100 Rotating Self-laminating Vinyl Wire Labels

Identify your wires from any angle with B-427 Rotating Vinyl Wire Labels. Simply print your labels and then peel, fold, and wrap – they’ll rotate freely for easy ID on even closed network connections.

Supplier:  Brady™ THTRO2954271.5

 View more versions of this product

Catalog No. 19129523


Only null left

Description

Description

  • Rotates so you can identify your labels from any angle
  • Simply wrap and apply – there is no adhesive residue or label parts to cling to your wires
  • Spins freely without anchors so these labels won't damage your Cat 5/6 and fiber optic cables
Specifications

Chemical Identifiers

ASTM D 1000; ASTM D 2979; ASTM G 155; UL Recognized
None
Matte
Non-sterile
BradyPrinter i5100; BradyPrinter i7100; THT 3 in. Core Printer
Rectangle
Permanent
White/Translucent
Self-laminating Vinyl
Wire and Cable Labeling
Overlaminate Included
-94°F to +158°F

Specifications

ASTM D 1000; ASTM D 2979; ASTM G 155; UL Recognized
None
Matte
Label
Wire and Cable Labeling
Overlaminate Included
1500/Pk.
1 x 2.75 in. (W x H)
10 to 6 AWG
Permanent
White/Translucent
Self-laminating Vinyl
Non-sterile
BradyPrinter i5100; BradyPrinter i7100; THT 3 in. Core Printer
2
Rectangle
-94°F to +158°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions