Learn More
Thermo Scientific Chemicals Bradykinin Acetate Salt
For cellular and molecular biology applications | 1120.21 g/mol
$383.07 - $778.49
Specifications
| Purification Method | HPLC |
|---|---|
| Content And Storage | -30°C to -10°C |
| Format | Powder |
| Molecular Weight (g/mol) | 1120.21 |
| Product Type | Bradykinin acetate salt |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ60345MC
|
Thermo Scientific Chemicals
J60345MC |
100 mg |
Each for $778.49
|
|
|||||
|
AAJ60345MB
|
Thermo Scientific Chemicals
J60345MB |
25 mg |
Each for $383.07
|
|
|||||
Description
Bradykinin Acetate elicits vasoconstriction, a systemic blood pressure reflex, a stimulation of respiration, and contraction of the nictitating membrane. Bradykinin plays an important role in the regulation of fluid and electrolyte balance, smooth muscle contraction, vasodilation and capillary permeability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| HPLC | |
| Powder | |
| Bradykinin acetate salt | |
| Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH |
| -30°C to -10°C | |
| 1120.21 | |
| RUO - research use only |
RUO – Research Use Only