missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bradykinin (2-9)

Catalog No. AAJ66218MB
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66218MB 25 mg
AAJ66218LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66218MB Supplier Thermo Scientific Chemicals Supplier No. J66218MB

For cellular and molecular biology applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Powder
Molecular Weight (g/mol) 904.02
Product Type Bradykinin (2-9)
Quantity 25 mg
Regulatory Status RUO - research use only
Sequence Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg

RUO – Research Use Only