Learn More
Brady™ Write-on Inspection Labels
Write-on Inspection Labels from Brady™ provide easy identification for calibration, testing, and repair of tools and equipment.
Supplier:  Brady™ 82588
 View more versions of this product
Catalog No. 19040749
Only null left
Description
Description
  • Polyester film with an acrylic pressure sensitive adhesive and a printable coating formulated specially for laser printers and toners
  • High resolution, good solvent and smudge resistance, moderate to high temperature performance
  • RoHS compliant to Pass 2011/65/EU
  • White is a UL recognized component and CSA accepted material when printed with designated laser printers
Specifications
Chemical Identifiers
Acrylic
Polyester
Used for safety, maintenance and general markings within a facility
1 Year
-4°F to 194°F
20 lb./in.
1.5 in.
Label
Non-sterile
Inkjet and laser printers
364/Pk.
-40°C to +90°C
0.0034 in.
38.1 mm
Specifications
Acrylic
Matte
Polyester
Non-sterile
Used for safety, maintenance and general markings within a facility
1 Year
-4°F to 194°F
20 lb./in.
0.625 x 1.5 in.
38.1 mm
15.875 mm
White, Blue
Label
Label
Tested, Date, Initials
Inkjet and laser printers
364/Pk.
-40°C to +90°C
0.0034 in.
1.5 in.
0.625 in.
Product Suggestions
Product Suggestions