Learn More
Brady™ Warning Message Tapes
Catalog No. 19103768
Catalog No. 19103768 Supplier Brady™ Supplier No. 58255
Used to warn against potential hazards or to caution against unsafe practices
  • Holds on almost any clean, dry surface
  • Made of B-950 vinyl with permanent adhesive
  • Colors sealed beneath protective layer of abrasion resistant clear vinyl
  • Can be used in toughest indoor conditions
Certifications
Meet OSHA specification 29 CFR 1910.145(c)(2) for Yellow and Black Caution Tape.
Specifications
Color Black, Yellow