missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Warning: Electrical Hazard... Signs

Catalog No. 19035919
Change view
Click to view available options
Material:
Fiberglass
Plastic
Polyester
Height (English):
7 in.
10 in.
Width (English):
10 in.
14 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English)
19035919 Polyester 10 in. 14 in.
19035471 Plastic 10 in. 14 in.
19035486 Fiberglass 10 in. 14 in.
19035657 Polyester 7 in. 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19035919 Supplier Brady™ Supplier No. 84198

Ensures safety at workplace

  • Black font on orange background

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145, PSTC-33,
Material Polyester
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label
Height (English) 10 in.
Max. Temperature (English) 230°F
Min. Temperature (English) -40°F
Thickness 0.006 to 0.01 in.
Width (English) 14 in.
Temperature Range (Metric) -40°C to 110°C
Temperature Range (English) -40°F to 230°F
Displayed Text Warning Electrical Hazard Authorized Personnel Only
Product Type Electrical Hazard Sign
Color Black on Orange
Show More Show Less