missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Warning: Electrical Hazard... Signs
Ensures safety at workplace
Supplier: Brady™ 84197
Description
- Black font on orange background
Chemical Identifiers
| 1 Ea. | |
| -40°F | |
| Black on Orange |
| Surface Mount | |
| Warning Electrical Hazard Authorized Personnel Only |
Specifications
| ANSI Z535, OSHA 1910.145, PSTC-33, | |
| 1 Ea. | |
| Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label | |
| 230°F | |
| 0.006 to 0.01 in. | |
| -40°C to 110°C | |
| Warning Electrical Hazard Authorized Personnel Only | |
| Black on Orange |
| Polyester | |
| Surface Mount | |
| 7 in. | |
| -40°F | |
| 10 in. | |
| -40°F to 230°F | |
| Electrical Hazard Sign |