Learn More
Brady™ Vinyl Single Footprints
Catalog No. 19103713
Change view
Click to view available options
Product Type:
Left Footprint
Right Footprint
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19103713 Right Footprint
19103712 Left Footprint
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19103713 Supplier Brady™ Supplier No. 121406
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vinyl Single Footprints are designed for easy hand application.
  • Made of durable B-7569 vinyl film with permanent acrylic adhesive that adheres to rough and highly textured surfaces
  • Excellent chemical, oil and water resistance
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Service temperature of -4 to 167°F
Specifications
Product Type Right Footprint
Color Yellow