missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ToughStripe™ Warning: Forklift Traffic with Pictogram Floor Safety Sign

Catalog No. 19150840
Change view
Click to view available options
Material:
Polyester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19150840 Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19150840 Supplier Brady™ Supplier No. 104512

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use these signs in important places to remind forklift drivers and other warehouse employees to stop or take established paths to avoid potential hazards or accidents.

  • ToughStripe floor signs feature durable floor marking tape with a high-performance adhesive
  • Sign reads WARNING FORKLIFT TRAFFIC (with picto and anti-skid areas)
  • Black on yellow
  • Diameter: 43.18cm (17 in.)
  • B-534 polyester — surface printed polyester with clear polyester overlaminate, anti-skid areas and rubber-based adhesive

  • Bright colors and bold text provide high visibility to critical messages
  • Designed to withstand harsh industrial environments

Specifications

Certifications/Compliance ANSI A1264.2-2006, OSHA 1910.22
Alternate Part Number B534FS 17 in. BLK/YEL WRNG FORKLIFT TRAFFIC
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Rubber
Description ToughStripe - Pre-printed Floor Signs
Diameter (English) 17 in.
Diameter (Metric) 431.80 mm
For Use With (Application) For communicating important safety and warning messages to employees
Max. Temperature (English) 130°F
Min. Temperature (English) 0°F
Thickness 0.01 in.
Temperature Range (Metric) -18°C to 54°C
Shape Round
Temperature Range (English) 0°F to 130 °F
Languages English
Displayed Text Warning Forklift Traffic
Product Type Floor Marking Sign
Color Black on Yellow
Show More Show Less