missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ToughStripe™ Die-Cut Floor Marking Letter K

Catalog No. p-6579016
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19108205 Black
19108206 White
19108207 Red
19108208 Green
19108209 Blue
19108210 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19108205 Supplier Brady™ Supplier No. 121751
Only null left

For marking aisle-ways, shipping and receiving docks, distribution, or other areas of your facility

  • Developed for superior durability, easy application, and painless removal
  • More visible than paint
  • Contains uppercase letter

Specifications

Adhesive Type Permanent Rubber-based
Color Black