missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ToughStripe™ Die-Cut Floor Marking Letter I

Catalog No. 19108198
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19108198 Yellow
19108193 Black
19108194 White
19108195 Red
19108196 Green
19108197 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19108198 Supplier Brady™ Supplier No. 121744

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For marking aisle-ways, shipping and receiving docks, distribution, or other areas of your facility

  • Developed for superior durability, easy application, and painless removal
  • More visible than paint
  • Contains uppercase letter

Specifications

Adhesive Type Permanent Rubber-based
Color Yellow