missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ToughStripe™ Die-Cut Floor Marking Letter G
Description
- Developed for superior durability, easy application, and painless removal
- More visible than paint
- Contains uppercase letter
Specifications
Specifications
| Adhesive Type | Permanent Rubber-based |
| Color | Black |