Learn More
Brady™ Striped Warning Tapes
Description
- Permanent adhesive stands up to tough indoor conditions
- Colors are sealed beneath protective layer of abrasion resistant clear vinyl
- Holds on to almost any clean, dry surface
Specifications
Specifications
| Adhesive Type | Rubber-based Pressure-Sensitive |
| Color | Red, White |