Learn More
Brady™ Striped Anti-Skid Tapes
Description
- Made of rugged B-916 adhesive-backed polyester tape
- Coated with thousands of particles of durable aluminum oxide grit
- Bonds permanently to clean, dry floors and withstands grease and oil
- Aids visibility of hazardous areas
Specifications
Specifications
| Dimensions (L x W) | 60 ft. x 4 in. |
| Color | Black, Yellow |