Learn More
Brady™ Striped Anti-Skid Tapes
Catalog No. 19103781
Catalog No. 19103781 Supplier Brady™ Supplier No. 78148
Only null left
For indoor or outdoor use
  • Made of rugged B-916 adhesive-backed polyester tape
  • Coated with thousands of particles of durable aluminum oxide grit
  • Bonds permanently to clean, dry floors and withstands grease and oil
  • Aids visibility of hazardous areas
Specifications
Dimensions (L x W) 60 ft. x 3 in.
Color Black, Yellow