missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Signs: Stop

Catalog No. 19150839
Change view
Click to view available options
Material:
Polyester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19150839 Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19150839 Supplier Brady™ Supplier No. 104511

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

STOP ToughStripe™ Floor Signs feature a highly visible, glossy appearance for communicating important safety messages and anti-skid sections to help prevent slips, trips and falls.

  • ToughStripe™ Floor Signs communicate important safety and warning messages to your employees in areas with foot traffic
  • Durable ToughStripe™ Anti-Skid Polyester (B-534) includes a rigid, clear overlaminate and is selectively covered by anti-skid tread
  • Highly visible, glossy material resists harsh chemicals and is easy to apply and remove
  • Floor signs alert workers of potential hazards, direct traffic, and enforce protective equipment requirements.

Specifications

Certifications/Compliance ANSI A1264.2-2006, OSHA 1910.22
Alternate Part Number B534FS 17 in. DIA RED,BLK/WHT STOP W/SYMBOL
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Rubber-based
Description ToughStripe - Pre-printed Floor Signs
Diameter (English) 17 in.
Diameter (Metric) 431.80 mm
Mounting Type Self-adhesive
For Use With (Application) For communicating important safety and warning messages to employees
Max. Temperature (English) 130°F
Min. Temperature (English) 0°F
Thickness 0.01 in.
Temperature Range (Metric) -18°C to 54°C
Shape Round
Temperature Range (English) 0°F to 130 °F
Languages English
Displayed Text Stop
Product Type Floor Marking Sign
Color Black on White
Show More Show Less