missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Signs: Production Area
Description
- ToughStripe™ Floor Signs communicate important safety and warning messages to your employees in areas with foot traffic
- Durable ToughStripe™ Anti-Skid Polyester (B-534) includes a rigid, clear overlaminate and is selectively covered by anti-skid tread
- Highly visible, glossy material resists harsh chemicals and is easy to apply and remove
- Floor signs alert workers of potential hazards, direct traffic, and enforce protective equipment requirements.
Specifications
Specifications
| Certifications/Compliance | ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145 |
| Finish | Gloss |
| Material | Polyester |
| Quantity | 1 Ea. |
| Adhesive Type | Rubber-based |
| Description | ToughStripe - Pre-printed Floor Signs |
| Diameter (English) | 17 in. |
| Diameter (Metric) | 431.80 mm |
| For Use With (Application) | Used to mark aisles, passageways, storage locations, point out safety equipment and exits and enforce protective equipment requirements |
| Max. Temperature (English) | 129.9°F |
| Show More |