missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Signs: EMERGENCY SHELTER FOR HAZARDOUS WEATHER CONDITIONS

Catalog No. 19106147
Change view
Click to view available options
Dimensions (L x W):
14 x 10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19106147 14 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19106147 Supplier Brady™ Supplier No. 127161

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EMERGENCY SHELTER FOR HAZARDOUS WEATHER CONDITIONS signs quickly identify severe weather shelter locations.

  • Non-Reflective Aluminum (B-555) is ideal for indoor and outdoor usage
  • Printed with durable inks and clear-coat protection for fade resistance
  • Withstands wind, rain, sunlight and high temperatures
  • Average outdoor durability 5-8 years

Specifications

Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS);NFPA 704;OSHA 1910.1200
Material Aluminum
Adhesive Type Non-adhesive
Mounting Type Corner Holes
Height (English) 14 in.
Width (Metric) 25.4 cm
Shape Rectangular
Languages English
Dimensions (L x W) 14 x 10 in.
Product Type Sign
Color Red on White
Show More Show Less