missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Signs: DANGER: OVERHEAD POWER LINES

Catalog No. 19109499
Change view
Click to view available options
Dimensions (L x W):
10 x 14 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19109499 10 x 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19109499 Supplier Brady™ Supplier No. 123614

missing translation for 'validMainframeMsg'

DANGER: OVERHEAD POWER LINES signs identify electrical hazard and high voltage areas.

  • Non-Reflective Aluminum (B-555) is ideal for indoor and outdoor usage
  • Printed with durable inks and clear-coat protection for fade resistance
  • Withstands wind, rain, sunlight and high temperatures
  • Average outdoor durability 5-8 years

Specifications

Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Material Aluminum
Adhesive Type Non-adhesive
Mounting Type Corner Holes
Height (English) 10 in.
Width (Metric) 35.56 cm
Shape Rectangular
Languages English
Dimensions (L x W) 10 x 14 in.
Product Type Electrical Hazard Sign
Color Black
Show More Show Less