Learn More
Brady™ Signs: Danger Hydrochloric Acid
Danger: Hydrochloric Acid signs are safety signs from Brady™ that are available in a variety of materials, allowing you to choose the material and size that best fits your application.
Supplier:  Brady™ 84408
 View more versions of this product
Catalog No. 19-035718
Only null left
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
Specifications
Specifications
ANSI Z535, OSHA 1910.145
1 Ea.
Surface Mount
10 in.
-40°F
14 in.
-40°C to 110°C
10 x 14 in.
Chemical and Hazardous Materials Sign
Polyester
Permanent Acrylic
Identification of fire alarms, extinguishers and emergency exits
230°F
0.006 to 0.01 in.
35.56 cm
-40°F to 230°F
DANGER Hydrochloric Acid
Red on White
Product Suggestions
Product Suggestions