missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Signs: DANGER: HOT (W/PICTO)

Catalog No. 19110834
Change view
Click to view available options
Dimensions (L x W):
7 x 10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19110834 7 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19110834 Supplier Brady™ Supplier No. 46643

DANGER: HOT (with PICTOGRAM) signs communicate warning messages about the potential dangers of operating machinery and equipment in a facility. Signs remind employees about safety, risks, proper operating instructions and more.

  • B-555 signs are designed for use in light duty industrial, utility, commercial and institutional environments
  • Non-Reflective Aluminum (B-555) is ideal for indoor and outdoor usage
  • Uses UV inks and UV clear coat protection for fade resistance
  • Withstands wind, rain, sunlight and high temperatures

Specifications

Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Material Aluminum
Adhesive Type Non-adhesive
Height (English) 7 in.
Dimensions (L x W) 7 x 10 in.
Color Red on White