missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Signs: ARC FLASH PROTECTION

Catalog No. 19103204
Change view
Click to view available options
Dimensions (L x W):
7 x 10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19103204 7 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19103204 Supplier Brady™ Supplier No. 102523

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Signs specify the personal protection equipment required for each arc flash category.

Post signs in work area and near electrical equipment to boost awareness and compliance.

Specifications

Material Polystyrene
Mounting Type Corner Holes
Thickness 0.0591 in.
Dimensions (L x W) 7 x 10 in.
Displayed Text ARC FLASH PROTECTION
Color Black