missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Sign: Caution: Corrosive Materials

Catalog No. 19036233
Change view
Click to view available options
Material:
Polyester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19036233 Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19036233 Supplier Brady™ Supplier No. 84297

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents. Caution: Corrosive Materials Sign B-302 is an high performance pressure sensitive safety sign.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “CAUTION: CORROSIVE MATERIALS WEAR REQUIRED PROTECTION”
  • Color: Black on yellow
  • Size: 17.78H x 25.4cmW (7 x 10 in.)
  • B-302 self-sticking polyester; surface printed polyester, polyester overlaminate and sensitive acrylic
  • Designed to withstand harsh industrial environments
  • Available in other sizes and materials

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Mounting Type Surface Mount
For Use With (Application) Identification of fire alarms, extinguishers and emergency exits
Height (English) 7 in.
Max. Temperature (English) 230°F
Min. Temperature (English) -40°F
Thickness 0.006 to 0.01 in.
Width (English) 10 in.
Width (Metric) 25.4 cm
Temperature Range (Metric) -40°C to 110°C
Shape Rectangular
Temperature Range (English) -40°F to 230°F
Languages English
Dimensions (L x W) 7 x 10 in.
Displayed Text CAUTION Corrosive Materials Wear Required Protection
Product Type Chemical and Hazardous Materials Sign
Color Black on Yellow
Show More Show Less