missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Self-Laminating Tags

Catalog No. 19016467
Change view
Click to view available options
Color:
Black, Red, White
Message:
DANGER DO NOT OPERATE Do Not Remove This Tag It is Here To Protect My Life. Name/Date Do Not Remove This Tag/Remarks
DANGER Do Not Remove This Tag/Remarks/See Other Side
Dimensions (L x W):
5.75 x 3 in.
7 x 4 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Message Dimensions (L x W)
19016467 Black, Red, White DANGER DO NOT OPERATE Do Not Remove This Tag It is Here To Protect My Life. Name/Date Do Not Remove This Tag/Remarks 5.75 x 3 in.
19040548 Black, Red, White DANGER Do Not Remove This Tag/Remarks/See Other Side 7 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19016467 Supplier Brady™ Supplier No. BRD65501
Only null left

Includes

10 Nylon ties

Self-Laminating Tags can be personalized by writing on special instructions, hazards or warnings.

  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates and other special instructions
  • Seal and protect your written message with a clear adhesive overlay
  • Withstands moisture, grease and dirt

Specifications

Quantity 10/Pk.
Certifications/Compliance OSHA
Color Black, Red, White
Diameter (English) Grommet 0.37 in.
Material Polyester
Message DANGER DO NOT OPERATE Do Not Remove This Tag It is Here To Protect My Life. Name/Date Do Not Remove This Tag/Remarks
For Use With (Application) For marking on-the-spot warning of dangerous conditions
Durability 3 to 5 Years
Height (English) 5.75 in.
Height (Metric) 14.6cm
Includes 10 nylon ties
Thickness 0.01 in.
Temperature Range (Metric) -40°C to 100°C
Temperature Range (English) -40°F to 212°F
Width (English) 3 in.
Width (Metric) 7.6 cm
Dimensions (L x W) 5.75 x 3 in.
Diameter (Metric) Hole 0.93 cm
Show More Show Less