missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SDS Elements Signs

Catalog No. 19000480
Change view
Click to view available options
Material:
Polyester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19000480 Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19000480 Supplier Brady™ Supplier No. 133172

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

To identify equipment, rooms and materials, which contain or are contaminated with hazardous agents

  • Available in fiberglass and self-sticking polyester material
  • Size: 25.4W x 35.5cm H(10 x 14 in.)

Specifications

Material Polyester
Quantity 1 Ea.
Description Self-sticking Polyester (B-302)
Mounting Type Wall Mount
Height (English) 14 in.
Height (Metric) 35.5 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Shape Rectangular
Languages English
Dimensions (L x W) 25.4 x 35.5cm
Displayed Text SDS Format
Product Type SDS Elements Sign
Color Black, Red on White
Show More Show Less