Learn More
Brady™ Quick-Align™ Symbols
For use with Blank Hazardous Communication Panels to identify hazardous materials in accordance with the NFPA system
Supplier:  Brady™ 58688
 View more versions of this product
Catalog No. 191302579
Only null left
Description
Description
  • Tough outdoor-grade self-sticking vinyl characters are black Gothic style (except for radiation trefoil)
  • Speciific hazard warnings (“Use no water”, radiation symbol, oxidizer, acid, alkalai, corrosive)
Always consult Material Safety Data Sheets for product-specific hazard information.
Order Info
Each package contains 10 identical characters.
Specifications
Chemical Identifiers
RoHS
10/Pk.
Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
-40°F
-40°C to 82°C
Black
Vinyl
Gothic
180°F
0.004 in.
-40°F to 180°F
Specifications
RoHS
10/Pk.
Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
180°F
0.004 in.
-40°F to 180°F
Quik-Align Kit
Vinyl
Gothic
4 in.
-40°F
-40°C to 82°C
COR
Black
Product Suggestions
Product Suggestions