missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Quick-Align™ Symbols

Catalog No. 191302578
Change view
Click to view available options
Displayed Text:
ACID
ALK
COR
OXY
Height (English):
4 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text Height (English)
191302578 ACID 4 in.
191302577 OXY 4 in.
191302580 ALK 4 in.
191302579 COR 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 191302578 Supplier Brady™ Supplier No. BRD58672

For use with Blank Hazardous Communication Panels to identify hazardous materials in accordance with the NFPA system

  • Tough outdoor-grade self-sticking vinyl characters are black Gothic style (except for radiation trefoil)
  • Speciific hazard warnings (“Use no water”, radiation symbol, oxidizer, acid, alkalai, corrosive)

Always consult Material Safety Data Sheets for product-specific hazard information.

Order Info

Each package contains 10 identical characters.

Specifications

Certifications/Compliance RoHS
Material Vinyl
Quantity 10/Pk.
Style Gothic
For Use With (Application) Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
Height (English) 4 in.
Max. Temperature (English) 180°F
Min. Temperature (English) -40°F
Thickness 0.004 in.
Temperature Range (Metric) -40°C to 82°C
Temperature Range (English) -40°F to 180°F
Displayed Text ACID
Product Type Hazardous Material Identifier
Color Black
Show More Show Less