missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Quick-Align™ Symbols
For use with Blank Hazardous Communication Panels to identify hazardous materials in accordance with the NFPA system
Supplier: Brady™ BRD58667
Description
- Tough outdoor-grade self-sticking vinyl characters are black Gothic style (except for radiation trefoil)
- Speciific hazard warnings (“Use no water”, radiation symbol, oxidizer, acid, alkalai, corrosive)
Always consult Material Safety Data Sheets for product-specific hazard information.
Order Info
Each package contains 10 identical characters.
Chemical Identifiers
| RoHS | |
| 10/Pk. | |
| Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs | |
| -40°F | |
| -40°C to 82°C | |
| Black |
| Vinyl | |
| Gothic | |
| 180°F | |
| 0.004 in. | |
| -40°F to 180°F |
Specifications
| RoHS | |
| 10/Pk. | |
| Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs | |
| 180°F | |
| 0.004 in. | |
| -40°F to 180°F | |
| Hazardous Material Identifier |
| Vinyl | |
| Gothic | |
| 4 in. | |
| -40°F | |
| -40°C to 82°C | |
| OXY | |
| Black |