missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Double RTK Compliance Center with MSDS Binders

Catalog No. 19103280
Catalog No. 19103280 Supplier Brady™ Supplier No. 2010DB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Double RTK Compliance Center with MSDS Binders help create awareness of company MSDS information.

  • This Right-to-Know Compliance Center helps you comply with OSHA requirements by keeping important Material Safety Data Sheets (MSDS) easily accessible
  • Bright red and yellow board makes the information center highly visible to employees
  • Helps prevent costly mistakes by providing potentially life-saving information to employees
  • Ideal for improving awareness of your company's safety information

Specifications

Product Type Double RTK Compliance Center with MSDS Binders
Includes Red polystyrene board, two binder racks and two MSDS binders
Width (English) 29 in.
Width (Metric) 73.6 cm
Height (English) 20 in.
Height (Metric) 50.8 cm
Depth (Metric) 11.4 cm
For Use With (Application) Wall mount