missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Notice: Do Not Dump Chemicals Down… Signs

Catalog No. 19035735
Change view
Click to view available options
Material:
Plastic
Polyester
Height (English):
7 in.
10 in.
Width (English):
10 in.
14 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English)
19035735 Polyester 10 in. 14 in.
19035496 Polyester 7 in. 10 in.
19035733 Plastic 7 in. 10 in.
19035734 Plastic 10 in. 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19035735 Supplier Brady™ Supplier No. 84464

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identifying equipment, rooms and materials that contain or are contaminated with hazardous agents

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “NOTICE DO NOT DUMP CHEMICALS DOWN THIS DRAIN”
  • Black/blue on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Certifications/Compliance ANSI Z535, OSHA 1910.145
    Material Polyester
    Quantity 1 Ea.
    Description Self-sticking Polyester (B-302)
    Mounting Type Surface Mount
    For Use With (Application) Identification of fire alarms, extinguishers and emergency exits
    Height (English) 10 in.
    Height (Metric) 25.4 cm
    Max. Temperature (English) 230°F
    Min. Temperature (English) -40°F
    Thickness 0.006 to 0.01 in.
    Width (English) 14 in.
    Width (Metric) 35.56 cm
    Temperature Range (Metric) -40°C to 110°C
    Shape Rectangular
    Temperature Range (English) -40°F to 230°F
    Languages English
    Displayed Text Notice Do Not Dump Chemicals Down This Drain
    Product Type Chemical and Hazardous Materials Sign
    Color Blue on White
    Show More Show Less