missing translation for 'onlineSavingsMsg'
Learn More

Brady™ MSDS Binder and User's Guide

Catalog No. 19016316
Change view
Click to view available options
Subject:
Safety and Training Materials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Subject
19016316 Safety and Training Materials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19016316 Supplier Brady™ Supplier No. BRD58678

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides a central location for MSDS information

Striped MSDS Binder

  • High-quality vinyl binder neatly organizes and protects MSDSs
  • Safety striped for high visibility
  • Inside pocket holds additional information
  • Spine is 2 in. and holds up to 400 MSDSs

MSDS User's Guide

  • Included with purchase of MSDS Binder
  • Features colorful illustrations and explanations
  • Ideal for use as employee handout to help met MSDS training requirements
  • Laminated and 3-hole punched

Specifications

Format Binder
Product Type MSDS Binder
Includes User Guide
For Use With (Application) Organizes and stores MSDS documents
Subject Safety and Training Materials