missing translation for 'onlineSavingsMsg'
Learn More

Brady™ MiniMark™ Industrial Label Printer Ribbon

Catalog No. 19018458
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
For Use With (Equipment):
Brady Label Printers
MiniMark Printer
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Equipment)
19018458 Black Brady Label Printers
19018454 White MiniMark Printer
19018453 Black MiniMark Printer
19018456 Blue MiniMark Printer
19018455 Red MiniMark Printer
19018457 Green MiniMark Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19018458 Supplier Brady™ Supplier No. 105031

Ribbons for use with Brady MiniMark printers

If printing on die cut labels, use Brady black wax ribbon part #105031

Compliance

Compliant with RoHS Directive

Specifications

Product Type Industrial Label Printer Ribbon
Color Black
For Use With (Equipment) Brady Label Printers
Length (English) 290 ft.
Width (English) 4.4 in.
Length (Metric) 88.39 m
Format Ribbon
Includes Self-laminating Vinyl labels and tamper evident metalized Polyester labels
Certifications/Compliance RoHS, 2011/65/EU
Quantity 2/Pk.